missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone H2B type 1-B Protein
A DNA sequence encoding the Homo sapiens (Human) Histone H2B type 1-B, was expressed in the hosts and tags indicated.
1644.75 NOK - 24050.00 NOK
Specifications
Accession Number | NP_066406.1 |
---|---|
Gene ID (Entrez) | 3018 |
Name | Histone H2B type 1-B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091230
|
enQuireBio™
QP6159-EC-100UG |
100 μg |
5325.00 NOK
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16011240
|
enQuireBio™
QP6159-EC-1MG |
1 mg |
24050.00 NOK
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16001240
|
enQuireBio™
QP6159-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021240
|
enQuireBio™
QP6159-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16031240
|
enQuireBio™
QP6159-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041240
|
enQuireBio™
QP6159-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_066406.1 | |
Histone H2B type 1-B Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3018 | |
Research Use Only | |
HIST1H2BB | |
Recombinant Protein | |
E. coli | |
PEPSKSAPAP KKGSKKAITK AQKKDGKKRK RSRKESYSIY VYKVLKQVHP DTGISSKAMG IMNSFVNDIF ERIAGEASRL AHYNKRSTIT SREIQTAVRL LLPGELAKHA VSEGTKAVTK YTSSK | |
Greater than 90% as determined by SDS-PAGE. |