missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ENPEP / Aminopeptidase A Protein

A DNA sequence encoding the Homo sapiens (Human) ENPEP / Aminopeptidase A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8812-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16016621

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001968.3
ENPEP / Aminopeptidase A Protein
Research Use Only
ENPEP
Recombinant Protein
E. coli
YFQGQVKPIA DSLGWNDAGD HVTKLLRSSV LGFACKMGDR EALNNASSLF EQWLNGTVSL PVNLRLLVYR YGMQNSGNEI SWNYTLEQYQ KTSLAQEKEK LLYGLASVKN VTLLSRYLDL LKDTNLIKTQ DVFTVIRYIS YNSYGKNMAW NWIQLNWDYL VNRYTLNNRN LGRIVTIAEP FNTELQLWQM ESFFAKYPQA GAGEKPREQV LETVKNNIEW LKQHRNTIRE W
Greater than 90% as determined by SDS-PAGE.
2028
200 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers