missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ENPEP / Aminopeptidase A Protein
A DNA sequence encoding the Homo sapiens (Human) ENPEP / Aminopeptidase A, was expressed in the hosts and tags indicated.
1644.75 NOK - 24050.00 NOK
Specifications
Accession Number | NP_001968.3 |
---|---|
Gene ID (Entrez) | 2028 |
Name | ENPEP / Aminopeptidase A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16086611
|
enQuireBio™
QP8812-EC-100UG |
100 μg |
5325.00 NOK
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16006621
|
enQuireBio™
QP8812-EC-1MG |
1 mg |
24050.00 NOK
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16096611
|
enQuireBio™
QP8812-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16016621
|
enQuireBio™
QP8812-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026621
|
enQuireBio™
QP8812-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036621
|
enQuireBio™
QP8812-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001968.3 | |
ENPEP / Aminopeptidase A Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
2028 | |
Research Use Only | |
ENPEP | |
Recombinant Protein | |
E. coli | |
YFQGQVKPIA DSLGWNDAGD HVTKLLRSSV LGFACKMGDR EALNNASSLF EQWLNGTVSL PVNLRLLVYR YGMQNSGNEI SWNYTLEQYQ KTSLAQEKEK LLYGLASVKN VTLLSRYLDL LKDTNLIKTQ DVFTVIRYIS YNSYGKNMAW NWIQLNWDYL VNRYTLNNRN LGRIVTIAEP FNTELQLWQM ESFFAKYPQA GAGEKPREQV LETVKNNIEW LKQHRNTIRE W | |
Greater than 90% as determined by SDS-PAGE. |