missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CXCL13 / BCA-1 / BLC Protein
A DNA sequence encoding the Homo sapiens (Human) CXCL13 / BCA-1 / BLC, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8533-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006410.1 | |
CXCL13 / BCA-1 / BLC Protein | |
Research Use Only | |
CXCL13 | |
Recombinant Protein | |
E. coli | |
VLEVYYTSLR CRCVQESSVF IPRRFIDRIQ ILPRGNGCPR KEIIVWKKNK SIVCVDPQAE WIQRMMEVLR KRS | |
Greater than 90% as determined by SDS-PAGE. |
10563 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |