missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CXCL13 / BCA-1 / BLC Protein
A DNA sequence encoding the Homo sapiens (Human) CXCL13 / BCA-1 / BLC, was expressed in the hosts and tags indicated.
1644.75 NOK - 19129.63 NOK
Specifications
Accession Number | NP_006410.1 |
---|---|
Gene ID (Entrez) | 10563 |
Name | CXCL13 / BCA-1 / BLC Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16085141
|
enQuireBio™
QP8533-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16025151
|
enQuireBio™
QP8533-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075141
|
enQuireBio™
QP8533-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16005151
|
enQuireBio™
QP8533-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16015151
|
enQuireBio™
QP8533-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095141
|
enQuireBio™
QP8533-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_006410.1 | |
CXCL13 / BCA-1 / BLC Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
10563 | |
Research Use Only | |
CXCL13 | |
Recombinant Protein | |
E. coli | |
VLEVYYTSLR CRCVQESSVF IPRRFIDRIQ ILPRGNGCPR KEIIVWKKNK SIVCVDPQAE WIQRMMEVLR KRS | |
Greater than 90% as determined by SDS-PAGE. |