missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Beta-defensin 130 Protein
A DNA sequence encoding the Homo sapiens (Human) Beta-defensin 130, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9781-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001032893.1 | |
Beta-defensin 130 Protein | |
Research Use Only | |
DEFB130 | |
Recombinant Protein | |
Yeast | |
GVIPGQKQCI ALKGVCRDKL CSTLDDTIGI CNEGKKCCRR WWILEPYPTP VPKGKSP | |
Greater than 90% as determined by SDS-PAGE. |
100133267 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |