missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Beta-defensin 130 Protein
A DNA sequence encoding the Homo sapiens (Human) Beta-defensin 130, was expressed in the hosts and tags indicated.
2967.00 NOK - 32744.50 NOK
Specifications
Accession Number | NP_001032893.1 |
---|---|
Gene ID (Entrez) | 100133267 |
Name | Beta-defensin 130 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16088701
|
enQuireBio™
QP9781-YE-100UG |
100 μg |
9275.00 NOK
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16028711
|
enQuireBio™
QP9781-YE-500UG |
500 μg |
26725.00 NOK
500µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16098701
|
enQuireBio™
QP9781-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018711
|
enQuireBio™
QP9781-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038711
|
enQuireBio™
QP9781-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008711
|
enQuireBio™
QP9781-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001032893.1 | |
Beta-defensin 130 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
100133267 | |
Research Use Only | |
DEFB130 | |
Recombinant Protein | |
Yeast | |
GVIPGQKQCI ALKGVCRDKL CSTLDDTIGI CNEGKKCCRR WWILEPYPTP VPKGKSP | |
Greater than 90% as determined by SDS-PAGE. |