missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ankyrin repeat and SOCS box protein 1
A DNA sequence encoding the Homo sapiens (Human) Ankyrin repeat and SOCS box protein 6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8093-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001189332.1 | |
Ankyrin repeat and SOCS box protein 1 | |
Research Use Only | |
ASB6 | |
Recombinant Protein | |
E. coli | |
MPFLHGFRRI IFEYQPLVDA ILGSLGIQDP ERQESLDRPS YVASEESRIL VLTELLERKA HSPFYQEGVS NALLKMAELG LTRAADVLLR HGANLNFEDP VTYYTALHIA VLRNQPDMVE LLVHHGADVN RRDREKLLCS MLWPAATGCR STILRTFVSY WKEGQTSRPP PKMGTQCSPA SSSCLVRPWE GTKRRPR | |
Greater than 90% as determined by SDS-PAGE. |
140459 | |
100 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |