missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ankyrin repeat and SOCS box protein 1
A DNA sequence encoding the Homo sapiens (Human) Ankyrin repeat and SOCS box protein 6, was expressed in the hosts and tags indicated.
1644.75 NOK - 24050.00 NOK
Specifications
Accession Number | NP_001189332.1 |
---|---|
Gene ID (Entrez) | 140459 |
Name | Ankyrin repeat and SOCS box protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16002631
|
enQuireBio™
QP8093-EC-100UG |
100 μg |
5325.00 NOK
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16022631
|
enQuireBio™
QP8093-EC-1MG |
1 mg |
24050.00 NOK
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16012631
|
enQuireBio™
QP8093-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16032631
|
enQuireBio™
QP8093-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16042631
|
enQuireBio™
QP8093-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16052631
|
enQuireBio™
QP8093-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001189332.1 | |
Ankyrin repeat and SOCS box protein 1 | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
140459 | |
Research Use Only | |
ASB6 | |
Recombinant Protein | |
E. coli | |
MPFLHGFRRI IFEYQPLVDA ILGSLGIQDP ERQESLDRPS YVASEESRIL VLTELLERKA HSPFYQEGVS NALLKMAELG LTRAADVLLR HGANLNFEDP VTYYTALHIA VLRNQPDMVE LLVHHGADVN RRDREKLLCS MLWPAATGCR STILRTFVSY WKEGQTSRPP PKMGTQCSPA SSSCLVRPWE GTKRRPR | |
Greater than 90% as determined by SDS-PAGE. |