missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 5-formyltetrahydrofolate cyclo-ligase Protein
A DNA sequence encoding the Homo sapiens (Human) 5-formyltetrahydrofolate cyclo-ligase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6392-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001186687.1 | |
5-formyltetrahydrofolate cyclo-ligase Protein | |
Research Use Only | |
MTHFS | |
Recombinant Protein | |
E. coli | |
AAAAVSSAKR SLRGELKQRL RAMSAEERLR QSRVLSQKVI AHSEYQKSKR ISIFLSMQDE IETEEIIKDI FQRGKICFIP RYRFQSNHMD MVRIESPEEI SLLPKTSWNI PQPGEGDVRE EALSTGGLDL IFMPGLGFDK HGNRLGRGKG YYDAYLKRCL QHQEVKPYTL ALAFKEQICL QVPVNENDMK VDEVLYEDSS TA | |
Greater than 90% as determined by SDS-PAGE. |
10588 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |