missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 5-formyltetrahydrofolate cyclo-ligase Protein
A DNA sequence encoding the Homo sapiens (Human) 5-formyltetrahydrofolate cyclo-ligase, was expressed in the hosts and tags indicated.
1644.75 NOK - 24050.00 NOK
Specifications
Accession Number | NP_001186687.1 |
---|---|
Gene ID (Entrez) | 10588 |
Name | 5-formyltetrahydrofolate cyclo-ligase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16082980
|
enQuireBio™
QP6392-EC-100UG |
100 μg |
5325.00 NOK
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16002990
|
enQuireBio™
QP6392-EC-1MG |
1 mg |
24050.00 NOK
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16092980
|
enQuireBio™
QP6392-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16012990
|
enQuireBio™
QP6392-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16022990
|
enQuireBio™
QP6392-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16032990
|
enQuireBio™
QP6392-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001186687.1 | |
5-formyltetrahydrofolate cyclo-ligase Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
10588 | |
Research Use Only | |
MTHFS | |
Recombinant Protein | |
E. coli | |
AAAAVSSAKR SLRGELKQRL RAMSAEERLR QSRVLSQKVI AHSEYQKSKR ISIFLSMQDE IETEEIIKDI FQRGKICFIP RYRFQSNHMD MVRIESPEEI SLLPKTSWNI PQPGEGDVRE EALSTGGLDL IFMPGLGFDK HGNRLGRGKG YYDAYLKRCL QHQEVKPYTL ALAFKEQICL QVPVNENDMK VDEVLYEDSS TA | |
Greater than 90% as determined by SDS-PAGE. |