missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | RCL1 |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Immunocytochemistry, Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16854464
|
Proteintech
15330-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16844464
|
Proteintech
15330-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RCL1, which does not have cyclase activity, plays a role in 40S- ribosomal-subunit biogenesis in the early pre-rRNA processing steps at sites A0, A1 and A2 that are required for proper maturation of the 18S RNA.Specifications
| RCL1 | |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| Q9JJT0, Q9Y2P8 | |
| 10171, 309301, 59028 | |
| RCL1 Fusion Protein Ag7058 | |
| Primary | |
| -20°C | |
| RCL1 |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| RCL1, RNAC, RPC2, RPCL1, RTC2 | |
| Rcl1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title