Learn More
RCL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15330-1-AP-20UL
 View more versions of this product
Product Code. 16854464
  • 1710.33 NOK / 20µL
Description
RCL1, which does not have cyclase activity, plays a role in 40S- ribosomal-subunit biogenesis in the early pre-rRNA processing steps at sites A0, A1 and A2 that are required for proper maturation of the 18S RNA.
Specifications
Specifications
RCL1
Polyclonal
Unconjugated
RCL1
RCL1, RNAC, RPC2, RPCL1, RTC2
Rabbit
Antigen Affinity Chromatography
RUO
10171, 309301, 59028
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunofluorescence
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9JJT0, Q9Y2P8
Rcl1
RCL1 Fusion Protein Ag7058
20 μL
Primary
Human, Rat, Mouse