missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBPJ Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14613-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Transcriptional regulator that plays a central role in Notch signaling, a signaling pathway involved in cell-cell communication that regulates a broad spectrum of cell-fate. determinations. Acts as a transcriptional repressor when it is not associated with Notch proteins. When associated with some Notch protein, it acts as a transcriptional activator that activates transcription of Notch target genes. Probably represses or activates transcription via the recruitment of chromatin remodeling complexes containing histone deacetylase or histone acetylase proteins, respectively. Specifically binds to the immunoglobulin kappa-type J segment recombination signal sequence.Specifications
RBPJ | |
Polyclonal | |
Unconjugated | |
RBPJ | |
CBF 1, CBF1, csl, IGKJRB, IGKJRB1, KBF2, NY-REN-30, RBP J, RBP J kappa, RBP JK, RBPJ, RBPJK, RBPSUH, SUH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19664, 3516 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P31266, Q06330 | |
RBPJ | |
RBPJ Fusion Protein Ag6206 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |