missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBPJ Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | RBPJ |
---|---|
Concentration | 0.21 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16803344
|
Proteintech
14613-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16893334
|
Proteintech
14613-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Transcriptional regulator that plays a central role in Notch signaling, a signaling pathway involved in cell-cell communication that regulates a broad spectrum of cell-fate. determinations. Acts as a transcriptional repressor when it is not associated with Notch proteins. When associated with some Notch protein, it acts as a transcriptional activator that activates transcription of Notch target genes. Probably represses or activates transcription via the recruitment of chromatin remodeling complexes containing histone deacetylase or histone acetylase proteins, respectively. Specifically binds to the immunoglobulin kappa-type J segment recombination signal sequence.Specifications
RBPJ | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P31266, Q06330 | |
19664, 3516 | |
RBPJ Fusion Protein Ag6206 | |
Primary | |
-20°C | |
RBPJ |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CBF 1, CBF1, csl, IGKJRB, IGKJRB1, KBF2, NY-REN-30, RBP J, RBP J kappa, RBP JK, RBPJ, RBPJK, RBPSUH, SUH | |
RBPJ | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |