missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM39 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21339-1-AP-20UL
This item is not returnable.
View return policy
Description
In infectious mononucleosis, anti-EBNA-1 antibodies are produced which cross-react with multiple normal human proteins. The cross-reactivity is due to anti-gly/ala antibodies that cross-react with host proteins containing configurations like those in the EBNA-1 repeat. One such antigen is RALY which is a member of the heterogeneous nuclear ribonucleoprotein gene family.Specifications
| RBM39 | |
| Polyclonal | |
| Unconjugated | |
| RBM39 | |
| CAPER, CAPERalpha, CC1.3, fSAP59, HCC1, RBM39, RNA binding motif protein 39, RNA binding protein 39, RNPC2, Splicing factor HCC1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 170791, 362251, 9584 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14498, Q8VH51 | |
| Rbm39 | |
| RBM39 Fusion Protein Ag15584 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction