missing translation for 'onlineSavingsMsg'
Learn More

RBM39 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. p-7210174 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16870945 20 μL 20µL
16860945 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16870945

Brand: Proteintech 213391AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

In infectious mononucleosis, anti-EBNA-1 antibodies are produced which cross-react with multiple normal human proteins. The cross-reactivity is due to anti-gly/ala antibodies that cross-react with host proteins containing configurations like those in the EBNA-1 repeat. One such antigen is RALY which is a member of the heterogeneous nuclear ribonucleoprotein gene family.

Specifications

Antigen RBM39
Applications Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RBM39
Gene Accession No. Q14498, Q8VH51
Gene Alias CAPER, CAPERalpha, CC1.3, fSAP59, HCC1, RBM39, RNA binding motif protein 39, RNA binding protein 39, RNPC2, Splicing factor HCC1
Gene Symbols Rbm39
Host Species Rabbit
Immunogen RBM39 Fusion Protein Ag15584
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170791, 362251, 9584
Target Species Human, Rat, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.