Learn More
GIBCO™ Rat (Sprague Dawley) Cytosol
Product Code. 10382980 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10382980 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10382980 Supplier GIBCO™ Supplier No. RTCYPL
  • ONLINE EXCLUSIVE PRICE
    478.00 NOK
    353.65 NOK / 1mL
    Save 124.35 NOK
    26% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Liver cytosolic fractions prepared from a pool of male Sprague Dawley rats. Total protein concentration: 20mg/mL
Liver cytosolic fractions prepared from a pool of male Sprague Dawley rats.
Total protein concentration: 20 mg/ml
We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.
For research use only. Not intended for human or animal therapeutic or diagnostic use.
Related Link
Learn more about our microsomes and subcellular fractions
TRUSTED_SUSTAINABILITY
Specifications
Donor Source Pooled
Gender Male
Physical Form Frozen
Tissue Liver
Quantity 1 mL
Purity Research Grade
Host Species Rat
Concentration 20 mg/mL
Product Type Rat Cytosol
Content And Storage Store between -60°C and -85°C.
Format Tube
Sub Cellular Localization Cytosol
Shipping Condition Dry Ice
Product Line Gibco