Learn More
Fisherbrand™ Radiation Beta Transport Boxes
Product Code. 12367127 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Height (Metric):
95 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
12367127 95 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12367127
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
Manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes; these serve as an ideal solution for short-term storage of radioactive waste or radioisotopes.
  • Combines a shield with a convenient contained work area and storage container
  • The hinged lid creates a 15° angled shield when fully open
  • The lid can be locked shut for safety while transporting the unit and its contents
  • Side handles are fitted for easy lifting and cushioned non-slip feet prevent slipping
  • Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
  • These serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Both the Beta and Gamma storage bins are available in five sizes, while the two largest bin models have wheels for easy transportation
  • Also available is our range of floor-standing and benchtop bins with anti-slip feet and hinged lids, which serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Optional heavy duty drawstring bags may also be purchased
Specifications
Depth (Metric) Exterior 400 mm
Depth (Metric) Interior 380 mm
Height (Metric) 95 mm
Height (Metric) Exterior 95 mm
Height (Metric) Interior 75 mm
Width (Metric) Exterior 315 mm
Width (Metric) Interior 295 mm
Product Type Radiation Transit Box