Learn More
Fisherbrand™ Radiation Safety Boxes
Code produit 12347117 Tous les produits Fisherbrand Traceable Produits
Modifier l'affichage
Click to view available options
Height (Metric):
130mm
Conditionnement:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Height (Metric) Conditionnement
12347117 130mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 12347117
Estimated Shipment: 26-04-2027
Ajouter au panier
Ajouter au panier
Radiation boxes, with hinged lid and central swivel covers, that are ideal for protection against Beta or Gamma radiation.
  • 15 with hinged lid and swivel cover for disposable of pipette tips 2L capacity 130 x 130 x 130mm
  • Beta – 2L – (13 x 13 x 13cm), use with bag CSR-BAG1
  • Radiation boxes, with hinged lid and central swivel covers, Beta Box/Gamma Box
  • These boxes have a central hole covered by a hinged lid, which reduces the exposure to radiation during pipetting procedures
  • Available for either gamma or beta radiation protection
  • Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts to hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
  • Also available is our range of floor-standing and benchtop bins with anti-slip feet and hinged lids, which serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Both the Beta and Gamma storage bins are available in five sizes, and the two largest bin models have wheels for easy transportation
  • Optional heavy duty drawstring bags may also be purchased
Spécification
Dimensions (L x W x H) 13 x 13 x 13cm
For Use With (Equipment) For use with bag CSR-BAG1
Height (Metric) 130mm
Width (Metric) 130mm
Capacity 2 L
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.