Learn More
Fisherbrand™ Radiation Block Beta Containers
Product Code. 12387197 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Height (Metric):
120mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
12387197 120mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12387197
Estimated Shipment: 30-03-2027
Add to basket
Add to basket
Ideal solution for storing radioactive materials, to hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes.
  • Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts to hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
  • Also available is our range of floor-standing and benchtop bins with anti-slip feet and hinged lids, which serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Both the Beta and Gamma storage bins are available in five sizes, and the two largest bin models have wheels for easy transportation
  • Optional heavy duty drawstring bags may also be purchased
  • Beta storage/transport block
  • For storage and transport solid acrylic for 24 x 1.5mL and 15 x 0.5mL tubes 150mm x 120mm x 70mm external Fisherbrand
Specifications
Dimensions (L x W x H) 70 x 150 x 120mm
Height (Metric) 120mm
Length (Metric) 70mm
Product Type Radiation storage container
Tube Size 24 x 1.5mL and 15 x 0.5mL
Width (Metric) 150mm