Learn More
Gilson™ Rack Heightener RH2
Product Code. 16646155 Shop All Gilson Products
Change view
Click to view available options
Length (Metric):
33 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
16646155 33 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16646155 Supplier Gilson™ Supplier No. 27595513
  • ONLINE EXCLUSIVE PRICE
    5910.00 NOK
    5178.17 NOK / Pack of 1
    Save 731.83 NOK
    12% Off
Rack Heightener RH2
  • Gilson
  • 33 mm
  • Rack Heightener RH2
  • 200 Series
Specifications
Description 200 Series
Length (Metric) 33 mm