Learn More
RAB11B Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28498-1-AP-20UL
 View more versions of this product
Product Code. 16886165
  • 1710.33 NOK / 20µL
Description
The RAB11B antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RAB11B. This antibody recognizes human, mouse antigen. The RAB11B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
RAB11B
Polyclonal
Unconjugated
20 μL
9230
IgG
Western Blot, ELISA
200 μg/mL
Rabbit
Primary
Human, Mouse