Learn More
Gilson™ Minipuls™ 3 R8 Medium-Flow Pump Head
Product Code. 10571283 Shop All Gilson Products
Change view
Click to view available options
Flow Rate:
0.3μL to 30 mL/min.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Unit Size
10571283 0.3μL to 30 mL/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10571283 Supplier Gilson™ Supplier No. F117608
  • ONLINE EXCLUSIVE PRICE
    31110.00 NOK
    22763.90 NOK / Pack of 1
    Save 8346.10 NOK
    26% Off
 Restricted Item
Estimated Shipment: 19-01-2027
For use with Peristaltic pump
  • Standard model
  • With 10 stainless steel rollers per head
  • Eight channels
TRUSTED_SUSTAINABILITY
Specifications
Flow Rate 0.3μL to 30 mL/min.
Model R8 medium flow pump head
No. of Channels 8
No. of Rollers 10
Roller Material Stainless Steel
Type Peristaltic Pump Head