Learn More
Gilson™ R1 Medium Flow Pump Head for Gilson™ Peristaltic Pump
Product Code. 10347472 Shop All Gilson Products
Change view
Click to view available options
Flow Rate:
0.3μL/min. to 30 mL/min.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Unit Size
10347472 0.3μL/min. to 30 mL/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10347472 Supplier Gilson™ Supplier No. F117604
  • ONLINE EXCLUSIVE PRICE
    10905.00 NOK
    9858.58 NOK / Pack of 1
    Save 1046.42 NOK
    9% Off
 Restricted Item
Estimated Shipment: 02-11-2026
Add channels to your MINIPULS™ 3 or Evolution and allow even greater flexibility in your laboratory.
MINIPULS 3 Pump Heads
Choose from these MINIPULS 3 pump heads:
  • R1—Single channel pump head
  • R4—Four channel pump head
  • R8—Eight channel pump head
  • R2—Two channel pump head
  • R2HF—Two channel high flow pump head
  • R4HF—Four channel high flow pump head
MINIPULS Evolution Pump Heads
Choose from these MINIPULS Evolution pump heads:
  • MF1—Pump head with one cartridge
  • MF4—Pump head with four cartridges
  • HF2—Pump head with two high flow cartridges
TRUSTED_SUSTAINABILITY
Specifications
Flow Rate 0.3μL/min. to 30 mL/min.
No. of Channels 1