Learn More
R&D Systems™ Proteome Profiler Human Cell Stress Array Kit
Product Code. 18007631
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. 18007631
Brand: R&D Systems™ ARY018
The Proteome Profiler Human Cell Stress Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.
The Proteome Profiler Human Cell Stress Array Kit contains 4 membranes - each spotted in duplicate with 26 different cell stress antibodies.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit