Learn More
R&D Systems™ Mouse Lgr5/GPR49 (NP_034325) VersaClone cDNA
Product Code. 18743739
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18743739 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18743739 Supplier R&D Systems™ Supplier No. RDC0888
  • ONLINE EXCLUSIVE PRICE
    8200.00 NOK
    7754.25 NOK / 10µg
    Save 445.75 NOK
    5% Off
 Restricted Item
Estimated Shipment: 20-10-2026
Leucine-rich Repeat Containing G Protein-coupled Receptor 5
The Mouse Lgr5/GPR49 (NP_034325) VersaClone cDNA clone contains the complete ORF for Lgr5/GPR49, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Lgr5/GPR49 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Specifications
Gene mLgr5
Accession Number NP_034325
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 2737bp
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans