Learn More
R&D Systems™ Mouse Frizzled-3 (NP_067433) VersaClone cDNA
Product Code. 18482130
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18482130 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18482130 Supplier R&D Systems™ Supplier No. RDC0118
  • ONLINE EXCLUSIVE PRICE
    8200.00 NOK
    7754.25 NOK / 10µg
    Save 445.75 NOK
    5% Off
 Restricted Item
Estimated Shipment: 20-10-2026
FZD3
The Mouse Frizzled-3 (NP_067433) VersaClone cDNA clone contains the complete ORF for Frizzled-3, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Frizzled-3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Specifications
Gene mFZD3
Accession Number NP_067433
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 2014bp
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans