Learn More
R&D Systems™ Human FPRL1/FPR2 (NP_001453) VersaClone cDNA
Product Code. 18492310
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18492310 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18492310 Supplier R&D Systems™ Supplier No. RDC0059
  • ONLINE EXCLUSIVE PRICE
    3530.00 NOK
    3336.90 NOK / 10µg
    Save 193.10 NOK
    5% Off
 Restricted Item
Estimated Shipment: 11-11-2026
Formyl Peptide Receptor-like 1
The Human FPRL1/FPR2 (NP_001453) VersaClone cDNA clone contains the complete ORF for FPRL1/FPR2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The FPRL1/FPR2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Specifications
Gene hFPRL1/FPR2
Accession Number NP_001453
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 1068bp
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans