Learn More
(R)-(-)-4-Phenyl-2-oxazolidinone, 98%
Product Code. 11436320
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11436320
Brand: Thermo Scientific Alfa Aesar H27171.06
Request Bulk or Custom Format
CAS: 90319-52-1 | C9H9NO2 | 163.176 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 90319-52-1
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00192393
InChI Key QDMNNMIOWVJVLY-QMMMGPOBSA-N
Synonym r---4-phenyl-2-oxazolidinone, r-4-phenyloxazolidin-2-one, r-4-phenyl-2-oxazolidinone, 4r-4-phenyl-1,3-oxazolidin-2-one, 4r-4-phenyloxazolidin-2-one, 2-oxazolidinone, 4-phenyl-, 4r, r-4-phenyl-oxazolidin-2-one, pubchem6077, ksc131o9d, 4 r-phenyl-2-oxazolidinone
PubChem CID 730425
IUPAC Name (4R)-4-phenyl-1,3-oxazolidin-2-one
SMILES C1C(NC(=O)O1)C2=CC=CC=C2
Specifications
Melting Point 129°C to 132°C
Quantity 5 g
Formula Weight 163.18
Optical Rotation −55° (c=2 in Chloroform)
Percent Purity 98%
Chemical Name or Material (R)-(-)-4-Phenyl-2-oxazolidinone