CAS RN 90319-52-1
CAS RN 90319-52-1
IUPAC Name:
(4R)-4-phenyl-1,3-oxazolidin-2-one
Synonyms:
(4R)-4-phenyl-1,3-oxazolidin-2-one
Molecular Weight (g/mol):
163.18
Molecular Formula:
C9H9NO2
InChi Key:
QDMNNMIOWVJVLY-QMMMGPOBSA-N
SMILES:
O=C1N[C@@H](CO1)C1=CC=CC=C1
1
–
1
of
1
results
1
(R)-(-)-4-Phenyl-2-oxazolidinone, 98%, Thermo Scientific Chemicals
CAS: 90319-52-1 Molecular Formula: C9H9NO2 Molecular Weight (g/mol): 163.176 MDL Number: MFCD00192393 InChI Key: QDMNNMIOWVJVLY-QMMMGPOBSA-N Synonym: r---4-phenyl-2-oxazolidinone,r-4-phenyloxazolidin-2-one,r-4-phenyl-2-oxazolidinone,4r-4-phenyl-1,3-oxazolidin-2-one,4r-4-phenyloxazolidin-2-one,2-oxazolidinone, 4-phenyl-, 4r,r-4-phenyl-oxazolidin-2-one,pubchem6077,ksc131o9d,4 r-phenyl-2-oxazolidinone PubChem CID: 730425 IUPAC Name: (4R)-4-phenyl-1,3-oxazolidin-2-one SMILES: C1C(NC(=O)O1)C2=CC=CC=C2