missing translation for 'onlineSavingsMsg'
Learn More

Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 11469480
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469480 1 g 1g
11459480 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469480 Supplier Thermo Scientific Alfa Aesar Supplier No. H50318.03
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid, is used as a chemicals, pharmaceutical intermediates, reagents. It is an important raw material and intermediate used in pharmaceuticals, agro based and dye stuff fields.

Solubility
Soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25940-35-6
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.14
MDL Number MFCD01975194
InChI Key HNYVPKNVKSTVJO-UHFFFAOYSA-N
Synonym pyrazolo 1,5-a pyrimidine-3-carboxylic acid, pyrazolo 1,5-a pyrimidine-3-carboxylicacid, 3-carboxypyrazolo 1,5-a pyrimidine, 8-hydropyrazolo 1,5-a pyrimidine-3-carboxylic acid, pubchem20936, acmc-1cbw6, 3-pyrazolo 1,5-a pyrimidinecarboxylic acid
PubChem CID 1910189
IUPAC Name pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
SMILES OC(=O)C1=C2N=CC=CN2N=C1

Specifications

Melting Point ∽180°C (decomposition)
Quantity 1 g
Solubility Information Soluble in water
Formula Weight 163.14
Percent Purity 97%
Chemical Name or Material Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.