accessibility menu, dialog, popup

UserName

NNOOHN

CAS RN 25940-35-6

IUPAC Name: pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
Synonyms: pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
Molecular Weight (g/mol): 163.14
Molecular Formula: C7H5N3O2
InChi Key: HNYVPKNVKSTVJO-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2N=CC=CN2N=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)