Learn More
Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid, 97%, Thermo Scientific™
Product Code. 11459480
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459480 250 mg 250mg
11469480 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11459480 Supplier Thermo Scientific Alfa Aesar Supplier No. H50318.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid, is used as a chemicals, pharmaceutical intermediates, reagents. It is an important raw material and intermediate used in pharmaceuticals, agro based and dye stuff fields.
Solubility
Soluble in water
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25940-35-6
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.14
MDL Number MFCD01975194
InChI Key HNYVPKNVKSTVJO-UHFFFAOYSA-N
Synonym pyrazolo 1,5-a pyrimidine-3-carboxylic acid, pyrazolo 1,5-a pyrimidine-3-carboxylicacid, 3-carboxypyrazolo 1,5-a pyrimidine, 8-hydropyrazolo 1,5-a pyrimidine-3-carboxylic acid, pubchem20936, acmc-1cbw6, 3-pyrazolo 1,5-a pyrimidinecarboxylic acid
PubChem CID 1910189
IUPAC Name pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
SMILES OC(=O)C1=C2N=CC=CN2N=C1
Specifications
Melting Point ∽180°C (decomposition)
Quantity 250 mg
Solubility Information Soluble in water
Formula Weight 163.14
Percent Purity 97%
Chemical Name or Material Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid