Learn More
Fisherbrand™ Push-in Cap Glass Specimen Vials
Product Code. 15314779 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 882
Product Code. Color unitSize
15314779 Clear Pack of 882
1 options
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 15314779
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Store and display specimens securely and easily. Fisherbrand™ Push-in Cap Glass Specimen Vials are available in a variety of sizes. Fisherbrand™ screw-in polyethylene caps are sold separately.
  • Manufactured from clear Type 1B neutral glass
  • Versatile capacity range from a variety of sizes
  • 1 dram equates to approximately 3.55mL (or 0.125 fl. oz.)
  • Suitable for media, diagnostic, storage, display, and sample collection applications
  • Fisherbrand™ Polyethylene Push-in Caps for Glass Specimen Vials sold separately
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Diameter (Metric) Outer 11 mm
Material Neutral Glass (Type 1B)
Volume (Metric) 1.75 mL
Closure Type Push-in Cap
Closure Size 8 mm
Height (Metric) 36 mm