missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPRS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13008-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Interacts with LAR-interacting protein LIP.1. The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth.Specifications
PTPRS | |
Polyclonal | |
Unconjugated | |
PTPRS | |
PTPRS, PTPSIGMA, R PTP S, R PTP sigma, rPTPsigma | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19280, 25529, 5802 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.6 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
B0V2N1, Q13332, Q64605 | |
PTPRS | |
PTPRS Fusion Protein Ag3691 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |