missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPRS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | PTPRS |
---|---|
Concentration | 0.6 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16801014
|
Proteintech
13008-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16891004
|
Proteintech
13008-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Interacts with LAR-interacting protein LIP.1. The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth.Specifications
PTPRS | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
B0V2N1, Q13332, Q64605 | |
19280, 25529, 5802 | |
PTPRS Fusion Protein Ag3691 | |
Primary | |
-20°C | |
PTPRS |
0.6 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
PTPRS, PTPSIGMA, R PTP S, R PTP sigma, rPTPsigma | |
PTPRS | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |