Learn More
Fisherbrand™ PTFE Wing Stir Bars
Product Code. 16265781 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric):
5.5 mm
8 mm
10 mm
13 mm
For Use With (Equipment):
11-12 mm diameter tubes
14-15 mm diameter tubes
6-7 mm diameter tubes
9-10 mm diameter tubes
Height (Metric):
9mm
10mm
11mm
16mm
20mm
Unit Size:
Pack of 10
Product Code. Diameter (Metric) For Use With (Equipment) Height (Metric) unitSize
16265781 13 mm 14-15 mm diameter tubes 20mm Pack of 10
16225781 8 mm 9-10 mm diameter tubes 11mm Pack of 10
16235781 5.5 mm 6-7 mm diameter tubes 9mm Pack of 10
16255781 13 mm 14-15 mm diameter tubes 10mm Pack of 10
16245781 10 mm 11-12 mm diameter tubes 16mm Pack of 10
5 options
Product Code. 16265781
Only null left
Add to basket
Add to basket
Provides stable stirring action. The PTFE wing stir bars are effective in small tapered and round bottom test, centrifuge, and sample tubes.
  • Stable, general-purpose magnetic stir bar
  • For effective stirring in small tapered and round bottom test, centrifuge, and sample tubes
  • Autoclavable
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core
For Use With (Equipment) 14-15 mm diameter tubes
Height (Metric) 20mm
Diameter (Metric) 13 mm
Shape Wing