Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
| Color | White |
| Description | PTFE encapsulated Alnico V magnet core |
| For Use With (Equipment) | 14-15 mm diameter tubes |
| Height (Metric) | 10mm |
| Diameter (Metric) | 13 mm |
| Shape | Wing |