Learn More
Fisherbrand™ PTFE Stir Bars with Removable Ring
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
12 mm
25 mm
38 mm
42 mm
51 mm
75 mm
Diameter (Metric):
8 mm
10 mm
12 mm
Height (Metric):
8mm
10mm
12mm
Unit Size:
Pack of 10
Pack of 5
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
| Color | White |
| Description | PTFE encapsulated Alnico V magnet core |
| Dimensions (L x W x H) | 51 x 8 x 8mm |
| For Use With (Equipment) | Containers & Vessels / Stirrer Hot Plates |
| Height (Metric) | 8mm |
| Includes | PTFE Ring |
| Diameter (Metric) | 8 mm |
| Length (Metric) | 51 mm |
| Show More |