Learn More
Fisherbrand™ PTFE Stir Bars with Removable Ring
Product Code. 16275751 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
12 mm
25 mm
38 mm
42 mm
51 mm
75 mm
Diameter (Metric):
8 mm
10 mm
12 mm
Height (Metric):
8mm
10mm
12mm
Unit Size:
Pack of 10
Pack of 5
Product Code. Length (Metric) Diameter (Metric) Height (Metric) unitSize
16275751 51 mm 10 mm 10mm Pack of 10
16265751 42 mm 10 mm 10mm Pack of 10
16245751 38 mm 10 mm 10mm Pack of 10
16295751 75 mm 12 mm 12mm Pack of 5
16225751 12 mm 8 mm 8mm Pack of 10
16235751 25 mm 8 mm 8mm Pack of 10
16255751 38 mm 8 mm 8mm Pack of 10
16285751 51 mm 8 mm 8mm Pack of 10
8 options
Product Code. 16275751
Only null left
Add to basket
Add to basket
Stir contents inside flasks and other vessels for complete mixing. Fisherbrand™ PTFE Stir Bars with Removable Ring are available in a number of sizes to accommodate both small and large mixing jobs.
  • Manufactured from PTFE with Alnico V magnetic core
  • Removable pivot ring
  • Autoclavable
  • PTFE Coating
  • Includes a certificate of conformance and certificate of analysis
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core
Dimensions (L x W x H) 51 x 10 x 10mm
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Height (Metric) 10mm
Includes PTFE Ring
Diameter (Metric) 10 mm
Length (Metric) 51 mm
Quantity 2/Pk.
Shape Cylinder