Specifications
| Color | White |
| Description | PTFE encapsulated Alnico V magnet core |
| For Use With (Application) | Removal of Stirrer Bars from vessels |
| For Use With (Equipment) | PTFE Stirrer Bars |
| Height (Metric) | 10mm |
| Diameter (Metric) | 10 mm |
| Length (Metric) | 350 mm |
| Quantity | 1/Ea. |