Learn More
Fisherbrand™ PTFE Stir Bar with Removable Ring
Product Code. 10466402 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W x H):
32 x 8 x 8mm
102 x 16 x 16mm
127 x 16 x 16mm
150 x 19 x 19mm
Unit Size:
Each
Pack of 10
Product Code. Dimensions (L x W x H) unitSize
10466402 150 x 19 x 19mm Each
10500482 102 x 16 x 16mm Each
10120002 127 x 16 x 16mm Each
12606326 32 x 8 x 8mm Pack of 10
4 options
Product Code. 10466402
Only null left
Add to basket
Add to basket
Provides stable stirring action. Fisherbrand™ PTFE Stir Bar with Removable Ring are autoclavable and include a pivot ring.
  • Guaranteed inert and leak-free
  • Isostatically molded in pure, FDA approved, PTFE
  • Individually polished to stay cleaner for longer
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core
Dimensions (L x W x H) 150 x 19 x 19mm
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Height (Metric) 19mm
Includes PTFE Ring
Diameter (Metric) 19 mm
Length (Metric) 150 mm
Quantity 1/Ea.
Shape Cylinder