Specifications
| Color | White |
| Description | Pure PTFE Magnet stir bar |
| Dimensions (L x W x H) | 30mm x 7mm |
| For Use With (Equipment) | Stirrers, hotplate stirrers |
| Width (Metric) | 7mm |
| Diameter (Metric) | 7 mm |
| Length (Metric) | 30 mm |
| Quantity | 1/Pk. |
| Shape | Cylinder |