Learn More
Fisherbrand™ PTFE Micro Magnetic Stir Bar
Product Code. 16245851 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
2 mm
3 mm
5 mm
6 mm
7 mm
8 mm
10 mm
13 mm
15 mm
20 mm
30 mm
Diameter (Metric):
1.5 mm
2 mm
3 mm
Unit Size:
Each
Pack of 10
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) unitSize
16245851 30 mm 3 mm Pack of 10
11898882 8 mm 1.5 mm Pack of 10
10652401 10 mm 3 mm Each
11868882 3 mm 3 mm Pack of 10
11808892 13 mm 3 mm Pack of 10
11878882 5 mm 2 mm Pack of 10
11828892 15 mm 1.5 mm Pack of 10
12656316 20 mm 3 mm Pack of 10
11888882 6 mm 3 mm Pack of 10
11858882 2 mm 2 mm Pack of 10
11818892 8 mm 3 mm Pack of 10
10180301 5 mm 2 mm Each
11888952 7 mm 2 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 16245851
Only null left
Add to basket
Edge
Add to basket
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
  • Supplied in reusable snap cap vials.
  • For stirring in test tubes, small size beakers, or other containers.
  • Autoclavable.
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Diameter (Metric) 3 mm
Length (Metric) 30 mm
Quantity 5/Pk.
Shape Cylinder
Type Stir Bar, Micro