Learn More
Hanna Instruments™ PTFE Membrane Dissolved Oxygen Probe
Product Code. p-8013382
Click to view available options
Length (Metric) Cable:
10 m
2 m
4 m
Unit Size:
Each
Product Code. 11517829
Brand: Hanna Instruments™ HI76407/10F
Specifications
Length (Metric) Cable 10 m
Probe Type Standard DO
For Use With (Application) For both laboratory and field applications.
Temperature Sensor Thermistor temperature sensor