Learn More
Fisherbrand™ PTFE Flexible Magnet Retriever, Flexible 330x10mm
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
330 mm
Unit Size:
Each
Specifications
Specifications
| Color | White |
| Description | PTFE Flexible Stir Bar Retriever |
| For Use With (Equipment) | For the removal of Stir Bars from vessels of all kinds |
| Diameter (Metric) | 10 mm |
| Length (Metric) | 330 mm |
| Material | PTFE with Alnico V Magnet Core |
| Quantity | 1 |