Learn More
Fisherbrand™ PTFE Centrifuge Test Tube with Closure
Product Code. 10581242 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
8 mL
13 mL
17 mL
45 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10581242 8 mL Each
10417172 13 mL Each
10679122 17 mL Each
15913077 45 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10581242
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Estimated Shipment: 04-01-2027
Add to basket
Add to basket
Guaranteed inert and crack-free. Fisherbrand™ PTFE Centrifuge Test Tube with Closure can be used up to 280°C.
  • Isostatically molded in pure PTFE
  • Heavy wall construction
Specifications
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Closure Material PTFE
Closure Color White
Color White
Capacity (Metric) 8 mL
Material PTFE
For Use With PTFE Test Tube Racks
No. per Pack 1
Autoclavable Autoclavable
Bottom Shape Round
Closure Type Screw Cap
Diameter (Metric) 12 mm
Graduated No
Product Type Centrifuge Test Tube