Description
- Totally inert with PTFE encapsulated viton neck seal
- Total Chemical Resistance
- High Purity
- Temperature Range -200°C to +280°C
- Non contaminating
- Easy to clean
Specifications
For Use With (Equipment) | Reagent Bottles |
Description | Fisherbrand™ PTFE Bottle Pourer |
Material | PTFE |
Quantity | 1 Ea. |
Thread Size (Metric) | GL 45 |