Learn More
Fisherbrand™ PTFE 4-Way Connector
Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Inner:
6 mm
8 mm
Overall Length (Metric):
44 mm
48 mm
Width (Metric):
44 mm
48 mm
Height (Metric):
14 mm
Unit Size:
Each
Specifications
Specifications
| Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 8 mm |
| Material | PTFE |
| Overall Length (Metric) | 48 mm |
| Min. Operating Temperature (Metric) | -200°C |
| Packaging Quantity | 1 Each |
| Width (Metric) | 48 mm |
| Height (Metric) | 14 mm |
| Color | White |
| Show More |